Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium chloride, anhydrous ≥99.0% (by argentometric titration), crystallised, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Aluminum chloride

06220-1KG 06220-100G 06220-2.5KG
BJ06220-100GEA 78.31 USD InStock
BJ06220-100G BJ06220-1KG BJ06220-2.5KG
Aluminium chloride, anhydrous ≥99.0% (by argentometric titration), crystallised, Puriss. p.a., Fluka™
Aluminium chloride


Formula: AlCl₃
MW: 133.34 g/mol
Melting Pt: 190 °C (2.500 hPa)
Density: 2.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8,II
Order Now
Specification Test Results
Assay 99.0 - 101.0 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 10 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Magnesium (Mg) Max. 10 ppm
Sodium (Na) Max. 10 ppm
Nickel (Ni) Max. 10 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 10 ppm
Sulfate (SO₄) Max. 100 ppm
Vapor Pressure 1 mmHg ( 100 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty