Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Ammonium hydrogen carbonate 99-101%, Puriss., meets analytical specification of BP, Ph. Eur., E503, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Ammonium bicarbonate

11213-6X1KG
BJ11213-1KGCS 1112.81 USD InStock
BJ11213-1KG
Ammonium hydrogen carbonate 99-101%, Puriss., meets analytical specification of BP, Ph. Eur., E503, Fluka™
Ammonium hydrogen carbonate


Formula: NH₄HCO₃
MW: 79.06 g/mol
Melting Pt: 60 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012138
CAS Number: 1066-33-7
EINECS: 213-911-5
Order Now
Specification Test Results
Assay 99.0 - 101.0 %
Non-volatile matter Max. 100 ppm
Arsenic (As) Max. 1 ppm
Iron (Fe) Max. 5 ppm
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 2 ppm
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 10 ppm
Sulfate (SO₄) Max. 30 ppm
Residual solvents complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty