Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium iodide ≥99.0% (by argentometric titration), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals

09874-100G 09874-500G
BJ09874-100GEA 284.54 USD InStock
BJ09874-100G BJ09874-500G
Ammonium iodide ≥99.0% (by argentometric titration), Puriss. p.a. ACS, Fluka™
Ammonium iodide


Formula: NH₄I
MW: 144.94 g/mol
Density: 2.510 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7
Order Now
Specification Test Results
Assay 99.0 - 100.0 %
pH (5 %, 20°C) 4.5 - 6.5
Colour of the solution complying
Clearness of the solution complying
Residue on ignition (SO₄) Max. 0.02 %
Water insoluble matter Max. 0.005 %
Barium (Ba) Max. 20 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 1 ppm
Iron (Fe) Max. 2 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 0.001 %
Chloride and bromide (as Cl) Max. 50 ppm
Total S (as SO₄) Max. 20 ppm
Phosphate (PO₄) Max. 10 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty