Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocresol green Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

32742-5G
BJ32742-5GEA 88.58 USD InStock
BJ32742-5G
Bromocresol green Reag. Ph. Eur., Fluka™
Bromocresol green


Formula: C₂₁H₁₄Br₄O₅S
MW: 698.02 g/mol
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/13/36/2301336.jpg
Order Now
Specification Test Results
pH Range 3.8 - 5.4, yellow to blue
Indicator solution complying
Change range complying
Test of sensitivity complying
Curve course (UV) complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty