Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Cesium chloride ≥99.9995% (metals basis), TraceSELECT™ for trace analysis, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Cesium Chloride

90033-25G 90033-100G
BJ90033-25GEA 173.1 USD InStock
BJ90033-25G BJ90033-100G
Cesium chloride ≥99.9995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Cesium chloride
TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions


Formula: CsCl
MW: 168.36 g/mol
Melting Pt: 642 °C
Density: 3.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Order Now
Specification Test Results
Assay Min. 99.5 %
Aluminium (Al) Max. 0.02 ppm
Arsenic (As) Max. 0.005 ppm
Barium (Ba) Max. 0.5 ppm
Beryllium (Be) Max. 0.005 ppm
Bismuth (Bi) Max. 0.05 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.02 ppm
Cerium (Ce) Max. 0.01 ppm
Cobalt (Co) Max. 0.005 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.005 ppm
Dysprosium (Dy) Max. 0.01 ppm
Erbium (Er) Max. 0.01 ppm
Europium (Eu) Max. 0.01 ppm
Iron (Fe) Max. 0.05 ppm
Gallium (Ga) Max. 0.005 ppm
Gadolinium (Gd) Max. 0.01 ppm
Mercury (Hg) Max. 5 ppb
Holmium (Ho) Max. 0.01 ppm
Indium (In) Max. 0.005 ppm
Potassium (K) Max. 1 ppm
Lanthanum (La) Max. 0.01 ppm
Lithium (Li) Max. 0.1 ppm
Lutetium (Lu) Max. 0.01 ppm
Magnesium (Mg) Max. 0.05 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 1 ppm
Neodymium (nd) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Praseodymium (Pr) Max. 0.01 ppm
Rubidium (Rb) Max. 2 ppm
Antimony (Sb) Max. 0.05 ppm
Scandium (Sc) Max. 0.01 ppm
Selenium (Se) Max. 0.05 ppm
Samarium (Sm) Max. 0.01 ppm
Strontium (Sr) Max. 0.1 ppm
Terbium (Tb) Max. 0.01 ppm
Thorium (Th) Max. 0.01 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.05 ppm
Thulium (Tm) Max. 0.01 ppm
Uran (U) Max. 0.01 ppm
Vanadium (V) Max. 0.01 ppm
Yttrium (Y) Max. 0.01 ppm
Ytterbium (Yb) Max. 0.01 ppm
Zinc (Zn) Max. 0.02 ppm
Total N Max. 10 ppm
Total S (as SO₄) Max. 20 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty