Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(I) oxide, red ≥86% (Cu basis), Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Cuprous oxideDicopper oxide

12841-500G 12841-6X2.5KG
BJ12841-2.5KGCS 2076.66 USD InStock
BJ12841-2.5KG BJ12841-500G
Copper(I) oxide, red ≥86% (Cu basis), Fluka™
Copper (I) oxide


Formula: Cu₂O
MW: 143.09 g/mol
Melting Pt: 1.230 °C
Density: 6 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010974
CAS Number: 1317-39-1
EINECS: 215-270-7
Order Now
Specification Test Results
Assay of Cu Min. 86 %
Silver (Ag) Max. 0.005 %
Iron (Fe) Max. 0.05 %
Nickel (Ni) Max. 0.01 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty