Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) nitrate hemipentahydrate ≥98% (≥26% Cu basis), Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Cupric Nitrate HydrateCupric nitrate hemipentahydrateCopper dinitrate hemipentahydrateCupric nitrate 2.5-hydrateCupric nitrate hemi(pentahydrate)

12837-1KG 12837-500G 12837-2.5KG
BJ12837-2.5KGEA 273.27 USD InStock
BJ12837-2.5KG BJ12837-1KG BJ12837-500G
Copper(II) nitrate hemipentahydrate ≥98% (≥26% Cu basis), Fluka™
Copper (II) nitrate hemipentahydrate


Formula: Cu(NO₃)₂·2.5H₂O
MW: 232.59 g/mol
Melting Pt: 114 °C
Density: 2.320 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD02253001
CAS Number: 19004-19-4
UN: 3085
ADR: 5.1,I
Order Now
Specification Test Results
Assay Min. 98 %
Assay of Cu Min. 26 %
Water insoluble matter Max. 0.003 %
Iron (Fe) Max. 0.005 %
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.01 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty