Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eriochrome blue black B indicator for complexometry (C.I. 14640), Fluka™

Supplier: Honeywell Research Chemicals
Warning

45540-25G
BJ45540-25GEA 27.06 USD InStock
BJ45540-25G
Eriochrome blue black B indicator for complexometry (C.I. 14640), Fluka™
Eriochrome Blue Black B


Formula: C₂₀H₁₃N₂NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00003934
CAS Number: 3564-14-5
EINECS: 222-639-6
Order Now
Specification Test Results
Appearance of the substance complying
Absorption maximum 523 - 533 nm
Absorbtion (1%/1cm) Min. 100
Suitability for Complexometry complying

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty