Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Ethylene glycol ≥99.5%, Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: 1,2-Ethanediol1,2-Dihydroxyethane1,2-Dihydroxy ethaneEthanediolGlycol alcoholDihydroxyethaneEthylene alcoholEthane-1,2-diolMonoethylene glycol2-HydroxyethanolGlycol

03750-250ML 03750-6X1L 03750-5L
BJ03750-250MLEA 48.64 USD InStock
BJ03750-250ML BJ03750-5L BJ03750-1L
Ethylene glycol ≥99.5%, Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Ethylene glycol


Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 197 °C
Melting Pt: -12 °C
Density: 1.11 g/cm³ (20 °C)
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/75/734375.jpg
Order Now
Specification Test Results
Refractive Index n20/D 1.431 (lit.)
Assay (GC) Min. 99.5 %
Identity (IR) complying
Boiling range 195 - 197 °C
Density (D 20/20) 1.113 - 1.115
Refractive index (n 20/D) 1.430 - 1.432
Water (Karl Fischer) Max. 0.1 %
Sulfated ash Max. 0.005 %
Free acid (as CH₃COOH) Max. 0.001 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.2 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.05 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 1 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.2 ppm
Chloride (Cl) Max. 0.2 ppm
Sulfate (SO₄) Max. 10 ppm
Formaldehyde Max. 50 ppm
KMnO₄ reducing matter (as O) Max. 0.0003 %
APHA Max. 25
Explosion Limit 15.3 %
Ignition Residue ≤0.005% (as SO₄)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty