Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrobromic acid ≥48%, Purum p.a.

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Hydrogen bromide

18730-1L 18730-250ML
BJ18730-250MLEA 54.48 USD InStock
BJ18730-250ML BJ18730-1L
Hydrobromic acid ≥48%, Purum p.a.
Hydrogen bromide
  • Products with a guarantee certificate and/or suitable for the stated analytical application.


Formula: HBr
MW: 80.91 g/mol
Boiling Pt: 126 °C (1.013 hPa)
Melting Pt: -11 °C
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II
Order Now
Specification Test Results
Assay 48.0 - 50.0 %
Refractive index (n 20/D) 1.440 - 1.443
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Sulfate and sulfite (as SO₄) Max. 50 ppm
Vapor Pressure 320 psi ( 21.1 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty