Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid fuming 36.5 - 38%, meets analytical specification of BP, Ph. Eur., NF, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Muriatic acid

07102-2.5L-GL
BJ07102-2.5LGLEA 68.89 USD InStock
BJ07102-2.5LGL
Hydrochloric acid fuming 36.5 - 38%, meets analytical specification of BP, Ph. Eur., NF, Fluka™
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 42 °C (1.013 hPa)
Melting Pt: -35 °C
Density: 1.190 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 36.5 - 38.0 %
Residue on ignition (SO₄) Max. 0.002 %
Non-volatile matter Max. 0.005 %
Arsenic (As) Max. 0.00005 %
Iron (Fe) Max. 0.0001 %
Free chlorine (Cl) Max. 1 ppm
Sulfate (SO₄) Max. 2 ppm
Sulfate (SO₄) complying
Sulfite (SO₃) Max. 0.0005 %
Sulfite (SO₃) complying
Free Bromine or Chlorine complying
Bromide or Iodide complying
Appearance of the substance complying
Appearance of the solution complying
Residual solvents complying
Vapor Density 1.3 (vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty