Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid fuming ≥37% ISO, Reag. Ph. Eur. for the determination of mercury (max. 0.005 ppm Hg), Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Muriatic acid

30719-4X2.5L
BJ30719-2.5LCS 308.85 USD InStock
BJ30719-2.5L
Hydrochloric acid fuming ≥37% ISO, Reag. Ph. Eur. for the determination of mercury (max. 0.005 ppm Hg), Fluka™
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 42 °C (1.013 hPa)
Melting Pt: -35 °C
Density: 1.190 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay Min. 37.0 %
Residue on ignition (SO₄) Max. 0.0005 %
Non-volatile matter Max. 0.005 %
Ammonium (NH₄) Max. 0.0001 %
Aluminium (Al) Max. 0.000005 %
Arsenic (As) Max. 0.000001 %
Barium (Ba) Max. 0.000002 %
Beryllium (Be) Max. 0.000002 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000001 %
Cobalt (Co) Max. 0.000001 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00002 %
Germanium (Ge) Max. 0.000005 %
Mercury (Hg) Max. 0.0000005 %
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000002 %
Strontium (Sr) Max. 0.000001 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000001 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Heavy metals (as Pb) Max. 2 ppm
Bromide (Br) Max. 0.005 %
Free chlorine (Cl) Max. 0.5 ppm
Phosphate (PO₄) Max. 0.00005 %
Sulfate (SO₄) Max. 1 ppm
Sulfite (SO₃) Max. 0.0001 %
Appearance of the solution complying
APHA Max. 10

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty