Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrofluoric acid ≥48%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

30107-500ML 30107-6X1L 30107-5L
BJ30107-500MLEA 98.19 USD InStock
BJ30107-500ML BJ30107-5L BJ30107-1L
Hydrofluoric acid ≥48%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Hydrofluoric acid


Formula: HF
MW: 20.01 g/mol
Boiling Pt: 105 °C (1.013 hPa)
Melting Pt: -35 °C
Density: 1.170 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II
Order Now
Specification Test Results
Assay Min. 48 %
Residue on ignition (SO₄) Max. 0.0005 %
Silver (Ag) Max. 0.000002 %
Aluminium (Al) Max. 0.000005 %
Arsenic (As) Max. 0.000005 %
Barium (Ba) Max. 0.00001 %
Beryllium (Be) Max. 0.000002 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000001 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00002 %
Germanium (Ge) Max. 0.000005 %
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000002 %
Magnesium (Mg) Max. 0.00002 %
Manganese (Mn) Max. 0.000005 %
Molybdenum (Mo) Max. 0.000005 %
Sodium (Na) Max. 0.00002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000005 %
Strontium (Sr) Max. 0.000002 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000005 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Heavy metals (as Pb) Max. 0.00005 %
Chloride (Cl) Max. 0.0001 %
Hexafluorosilicic acid (H₂SiF₆) Max. 0.005 %
Phosphate (PO₄) Max. 0.00005 %
Sulfate (SO₄) Max. 0.0002 %
Sulfite (SO₃) Max. 0.0002 %
Vapor Density 1.27 (vs air)
Appearance (color) complying
Appearance (form) complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty