Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium nitrate ≥99.5%, Reagent Grade

Supplier: Honeywell Research Chemicals
Warning

227986-1KG
BJ227986-1KGEA 253.65 USD InStock
BJ227986-1KG
Lithium nitrate ≥99.5%, Reagent Grade
Lithium nitrate


Formula: LiNO₃
MW: 68.95 g/mol
Boiling Pt: > 450 °C (dec.)
Melting Pt: 264 °C (1.013 hPa)
Density: 2.38 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011094
CAS Number: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1,III
Order Now
Specification Test Results
Identity (IR) complying
Assay of Li 9 - 11 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty