Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium oxide light ≥99%, tested according to Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Magnesia usta

63096-500G
BJ63096-500GEA 192.61 USD InStock
BJ63096-500G
Magnesium oxide light ≥99%, tested according to Ph. Eur., Fluka™
Magnesium oxide


Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3.600 °C (1.013 hPa)
Melting Pt: 2.852 °C
Density: 3.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Order Now
Specification Test Results
Identity B complying
Assay (calculated to ignited substance) 98.0 - 100.5 %
Appearance of the solution complying
Loss on ignition Max. 8.0 %
In water soluble matter Max. 2.0 %
Insoluble in acetic acid Max. 0.1 %
Arsenic (As) Max. 4 ppm
Calcium (Ca) Max. 1.5 %
Iron (Fe) Max. 0.1 %
Heavy metals (as Pb) Max. 30 ppm
Chloride (Cl) Max. 0.15 %
Sulfate (SO₄) Max. 1.0 %
Bulk Density Max. 0.15 g/ml
Residual solvents complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty