Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium perchlorate ≥85% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Anhydrone

222283-500G 222283-5G 222283-100G
BJ222283-100GEA 105.92 USD InStock
BJ222283-100G BJ222283-5G BJ222283-500G
Magnesium perchlorate ≥85% ACS, Fluka™
Magnesium perchlorate


Formula: Cl₂MgO₈
MW: 223.21 g/mol
Melting Pt: 251 °C (dec.)
Density: 2.21 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011102
CAS Number: 10034-81-8
UN: 1475
ADR: 5.1,II
Order Now
Specification Test Results
Identity (IR) complying
Assay of Mg Min. 10.0 %
Loss on drying (190°C) Max. 8 %
Titrable Acid Max. 0.005 meq/g
Titrable Base Max. 0.025 meq/g
Water absorbing capacity Min. 25 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty