Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium sulfate, anhydrous ≥97%, Reagent Grade

Supplier: Honeywell Research Chemicals
Synonyms: Magnesium(II) sulfateBitter saltSulfuric acid magnesium salt (1:1)Epsom salt

208094-2.5KG
BJ208094-2.5KGEA 352 USD InStock
BJ208094-2.5KG
Magnesium sulfate, anhydrous ≥97%, Reagent Grade
Magnesium sulphate


Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1.130 °C (dec.)
Density: 2.7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/17/16640617.jpg
Order Now
Specification Test Results
Assay of Mg 19.6 - 20.8 %
Vapor Pressure <0.1 mmHg ( 20 °C)
Vapor Density <0.01 (vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty