Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(II) chloride ≥99.5% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Mercury dichlorideMercuric chlorideMercury Chloride

215465-500G
BJ215465-500GEA 581.83 USD InStock
BJ215465-500G
Mercury(II) chloride ≥99.5% ACS, Fluka™
Mercury (II) chloride


Formula: HgCl₂
MW: 271.5 g/mol
Boiling Pt: 302 °C
Melting Pt: 276 °C
Density: 5.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011041
CAS Number: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1,II
Order Now
Specification Test Results
Assay Min. 99.5 %
Residue on ignition a. reducing with HCOOH (SO₄)* Max. 0.02 %
Iron (Fe) Max. 0.002 %
In ether insoluble matter complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty