Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitric acid 64 - 66%, Puriss., Fluka™

Supplier: Honeywell Research Chemicals
Danger

07006-2.5L 07006-1L
BJ07006-2.5LEA 114.19 USD InStock
BJ07006-2.5L BJ07006-1L
Nitric acid 64 - 66%, Puriss., Fluka™
Nitric acid


Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 122 °C (1.013 hPa)
Melting Pt: -31 °C
Density: 1.400-1.480 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8,II
Order Now
Specification Test Results
Assay 64 - 66 %
Residue on ignition (SO₄) Max. 0.0005 %
Arsenic (As) Max. 0.0001 %
Calcium (Ca) Max. 0.0005 %
Iron (Fe) Max. 0.0001 %
Vapor Pressure 8 mmHg ( 20 °C)
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.0001 %
Sulfate (SO₄) Max. 0.0002 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty