Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boric acid ≥99.5%, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acidOrthoboric acid

B0252-2.5KG
BJB0252-2.5KGEA 153.24 USD InStock
BJB0252-2.5KG
Boric acid ≥99.5%, Reagent Grade
ortho-Boric acid


Formula: B(OH)₃
MW: 61.83 g/mol
Melting Pt: 171 °C
Density: 1.510 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg
Order Now
Specification Test Results
Assay (alkalimetric) Min. 99.5 %
Colour of the solution complying
Clearness of the solution complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty