Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydrogen sulfate ≥99%, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Potassium bisulphatePotassium bisulfate

31260-500G 31260-6X1KG
BJ31260-500GEA 82.55 USD InStock
BJ31260-500G BJ31260-1KG
Potassium hydrogen sulfate ≥99%, Puriss. p.a., Fluka™
Potassium hydrogen sulphate


Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 200 °C
Density: 2.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Order Now
Specification Test Results
Assay Min. 99 %
Aluminium (Al) Max. 0.002 %
Arsenic (As) Max. 0.00005 %
Calcium (Ca) Max. 0.002 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Magnesium (Mg) Max. 0.0005 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Chloride (Cl) Max. 0.0005 %
Phosphate (PO₄) Max. 0.0005 %
Total N Max. 0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty