Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium periodate ≥99.8% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Potassium metaperiodatePotassium tetroxoiodate (VII)

210056-100G
BJ210056-100GEA 217.43 USD InStock
BJ210056-100G
Potassium periodate ≥99.8% ACS, Fluka™
Potassium periodate


Formula: KlO₄
MW: 230 g/mol
Melting Pt: 582 °C
Density: 3.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011407
CAS Number: 7790-21-8
EINECS: 232-196-0
UN: 3085
ADR: 5.1,I
Order Now
Specification Test Results
Assay (excluding dried substance) 99.8 - 100.3 %
Manganese (Mn) Max. 1 ppm
Other halogens (as Cl) Max. 0.01 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty