Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium thiocyanate ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Potassium rhodanide

207799-2.5KG 207799-100G
BJ207799-100GEA 105.06 USD InStock
BJ207799-100G BJ207799-2.5KG
Potassium thiocyanate ≥99.0% ACS, Fluka™
Potassium thiocyanate


Formula: KSCN
MW: 97.18 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: 172 °C
Density: 1.89 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Order Now
Specification Test Results
Assay Min. 98.5 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 5.3 - 8.7
Ammonium (NH₄) Max. 3 ppm
Iron (Fe) Max. 2 ppm
Sodium (Na) Max. 0.005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.005 %
Matter consuming I (as I) complying
IR spectrum complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty