Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Reinecke's salt ≥93.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

183687-25G 183687-100G
BJ183687-100GEA 609.19 USD InStock
BJ183687-100G BJ183687-25G
Reinecke's salt ≥93.0% ACS, Fluka™
Reinecke's salt


Formula: C₄H₁₀CrN₇S₄
Melting Pt: 268-272 °C
Storage Temperature: Ambient
EINECS: 237-003-3
Order Now
Specification Test Results
Assay Min. 93.0 %
Insoluble in HCl Max. 0.05 %
Test of sensitivity complying

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty