Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate 0.01 M AgNO3 (0.01N) ≥99.5% volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

34294-1L
BJ34294-1LEA 210.36 USD InStock
BJ34294-1L
Silver nitrate 0.01 M AgNO3 (0.01N) ≥99.5% volumetric solution, Fluka™
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Factor 0.995 - 1.005

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty