Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium acetate ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Acetic acid sodium salt

241245-5G 241245-2.5KG 241245-500G 241245-1KG 241245-100G
BJ241245-500GEA 103.6 USD InStock
BJ241245-500G BJ241245-100G BJ241245-5G BJ241245-1KG BJ241245-2.5KG
Sodium acetate ≥99.0% ACS, Fluka™
Sodium acetate


Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: 324 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
Order Now
Specification Test Results
Assay (HClO₄-titr.) Min. 99.0 %
Water insoluble matter Max. 0.01 %
Loss on drying (120°C) Max. 1.0 %
pH (5 %, 25 °C) 7.0 - 9.2
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 0.001 %
Magnesium (Mg) Max. 0.002 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.002 %
Phosphate (PO₄) Max. 0.001 %
Sulfate (SO₄) Max. 0.003 %
IR spectrum complying
Form solid
pKa  4.76 (acetic acid)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty