Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium ammonium hydrogen phosphate tetrahydrate ≥99.0% (by non aqueous titration), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Sodium ammonium phosphate tetrahydrateAmmonium sodium phosphate tetrahydrate

71283-1KG 71283-250G
BJ71283-1KGEA 184.58 USD InStock
BJ71283-1KG BJ71283-250G
Sodium ammonium hydrogen phosphate tetrahydrate ≥99.0% (by non aqueous titration), Puriss. p.a., Fluka™
Sodium ammonium hydrogen phosphate tetrahydrate


Formula: NaNH₄HPO₄·4H₂O
MW: 209.07 g/mol
Density: 1.544 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066970
CAS Number: 7783-13-3
Order Now
Specification Test Results
Assay 99.0 - 101.0 %
Silver (Ag) Max. 5 ppm
Aluminium (Al) Max. 5 ppm
Arsenic (As) Max. 0.5 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 10 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Thallium (Tl) Max. 10 ppm
Zinc (Zn) Max. 5 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty