Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 0.1 M NaCl (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: HaliteSea saltSaline

35275-1L
BJ35275-1LEA 58.01 USD InStock
BJ35275-1L
Sodium chloride 0.1 M NaCl (0.1N) volumetric solution, Fluka™
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Density: 1.000 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-791-2
Order Now
Specification Test Results
Factor 0.999 - 1.001

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty