Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride ≥99.5% for HPLC, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: HaliteSea saltSaline

73575-250G
BJ73575-250GEA 390.54 USD InStock
BJ73575-250G
Sodium chloride ≥99.5% for HPLC, Fluka™
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1.465 °C (1.013 hPa)
Melting Pt: 800 °C
Density: 2.160 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Order Now
Specification Test Results
Assay 99.5 - 100.5 %
pH (10 %, 20°C) 5.0 - 8.0
Absorbance at 220 nm Max. 0.05
Absorbance at 230 nm Max. 0.03
Absorbance at 240 nm Max. 0.02
Absorbance at 260 nm Max. 0.01
Absorbance 500 nm Max. 0.01
Appearance of the solution (c=10%) complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty