Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dichromate dihydrate ≥99%, Purum, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium bichromate

12266-5KG 12266-1KG
BJ12266-1KGEA 304.26 USD InStock
BJ12266-1KG BJ12266-5KG
Sodium dichromate dihydrate ≥99%, Purum, Fluka™
Sodium dichromate dihydrate


Formula: Na₂Cr₂O₇·2H₂O
MW: 298.00 g/mol (261.97 anhy)
Boiling Pt: Decomposes on heating
Melting Pt: 357 °C (anhydr.)
Density: 2.52 g/cm³ (13 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149166
CAS Number: 7789-12-0
EINECS: 234-190-3
UN: 3086
ADR: 6.1,I
Order Now
Specification Test Results
Assay Min. 99 %
Chloride (Cl) Max. 0.05 %
Sulfate (SO₄) Max. 0.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty