Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium sulfate decahydrate ≥99.0% (by titrimetric analysis on dry substance), crystallised, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals

71970-1KG
BJ71970-1KGEA 84.36 USD InStock
BJ71970-1KG
Sodium sulfate decahydrate ≥99.0% (by titrimetric analysis on dry substance), crystallised, Puriss. p.a., Fluka™
Sodium sulphate decahydrate


Formula: Na₂SO₄·10H₂O
MW: 322.2 g/mol
Melting Pt: 32.4 °C
Density: 1.46 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149189
CAS Number: 7727-73-3
Order Now
Specification Test Results
Assay (calculated to the dried substance) 99.0 - 101.0 %
pH (5 %, 20°C) 5.0 - 8.0
Appearance of the solution complying
Loss on drying (130°C) 53 - 57 %
Calcium (Ca) Max. 20 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 20 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 5 ppm
Phosphate (PO₄) Max. 10 ppm
Total N Max. 5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty