Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tungstate dihydrate ≥99%, Purum, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Sodium tungsten oxide dihydrateTungstic acid sodium salt dihydrateSodium wolframate dihydrate

14304-6X500G
BJ14304-500GCS 2223.73 USD InStock
BJ14304-500G
Sodium tungstate dihydrate ≥99%, Purum, Fluka™
Sodium wolframate dihydrate


Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Melting Pt: 698 °C
Density: 4.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2
Order Now
Specification Test Results
Assay Min. 99 %
Free alkali (as NaOH) Max. 0.1 %
Iron (Fe) Max. 0.002 %
Molybdenum (Mo) Max. 0.01 %
Chloride (Cl) Max. 0.01 %
Sulfate (SO₄) Max. 0.02 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty