Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 95.0-98.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

258105-500ML 258105-100ML 258105-1L-PC 258105-2.5L 258105-1L 258105-500ML
BJ258105-500MLEA 93.3 USD InStock
BJ258105-500ML BJ258105-100ML BJ258105-1L-PC BJ258105-2.5L BJ258105500MLP BJ258105-1L
Sulfuric acid 95.0-98.0% ACS, Fluka™
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 310 °C (1.013 hPa)
Melting Pt: -14 to -10 °C
Density: 1.840 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Vapor Density <0.3 (25 °C, vs air)
Assay 95.0 - 98.0 %
Density (D 20/4) 1.825 - 1.836
Residue on ignition Max. 5 ppm
Ammonium (NH₄) Max. 2 ppm
Arsenic (As) Max. 0.01 ppm
Iron (Fe) Max. 0.2 ppm
Mercury (Hg) Max. 0.005 ppm
Heavy metals (as Pb) Max. 1 ppm
Chloride (Cl) Max. 0.2 ppm
Nitrate (NO₃) Max. 0.5 ppm
KMnO₄ reducing matter (as SO₂) Max. 2 ppm
APHA Max. 10
Appearance complying
Vapor Pressure 1 mmHg ( 146 °C)
Appearance Free from suspended or insoluble matter

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty