Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

Zinc chloride 98-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Zinc (II) chlorideButter of zinc

14422-500G 14422-25KG 14422-6X1KG
BJ14422-500GEA 148.98 USD InStock
BJ14422-500G BJ14422-25KG BJ14422-1KG
Zinc chloride 98-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Zinc chloride


Formula: ZnCl₂
MW: 136.3 g/mol
Melting Pt: 287-304 °C (1.013 hPa)
Density: 2.93 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Order Now
Specification Test Results
Assay 98.0 - 100.5 %
Oxide chloride (as ZnO) complying
pH (10 %, 20°C) 4.6 - 5.5
Ammonium (NH₄) Max. 300 ppm
Ammonium (NH₄) complying
Arsenic (As) Max. 0.0002 %
Calcium (Ca) Max. 0.01 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.02 %
Magnesium (Mg) Max. 0.01 %
Sodium (Na) Max. 0.01 %
Lead (Pb) Max. 0.005 %
Total N Max. 0.005 %
Sulfate (SO₄) Max. 200 ppm
Vapor Pressure 1 mmHg ( 428 °C)
Al, Ca, Fe, Mg and heavy metals complying
Alkali and alkaline earth Max. 1.0 %
Residual solvents complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty