Would you like to log in again?
SP Wilmad-LabGlass Low Form Weighing Bottles with Standard Taper Stopper, SP Industries
Supplier: SP Industries
LG-3830-102 LG-3830-104 LG-3830-100
76323-436EA 81.87 USD InStock
76323-436 76323-434 76323-432
SP Wilmad-LabGlass Low Form Weighing Bottles with Standard Taper Stopper, SP Industries
Bottles Weighing Bottles
Ideal for precise weighing of substances, this low form bottle has a wide opening for easy addition and removal of samples.
  • Standard taper pennyhead stopper with closed bottom forms a secure seal
  • Used in desiccators to achieve absolute sample dryness
  • Light weight allows simple tare settings
  • Ideal for hygroscopic samples
ASTM Type 1, Class A Borosilicate glass construction.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty