Would you like to log in again?
SP Wilmad-LabGlass Parr Weighing Bottle with Flat Top Cap, SP Industries
Supplier: SP Industries
LG-3850-100
76323-438EA 38.94 USD InStock
76323-438
SP Wilmad-LabGlass Parr Weighing Bottle with Flat Top Cap, SP Industries
Bottles Weighing Bottles
Created specifically for small samples, the Parr weighing bottle has an external ground joint for sample transfer without seal interference.
  • Flat top permits stacking for increased desiccator capacity
  • Light weight allows simple tare settings
  • Ideal for hygroscopic samples
ASTM Type 1, Class A Borosilicate glass construction.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty