Would you like to log in again?
SP Wilmad-LabGlass Tall Form Weighing Bottles with Flat Top Cap, SP Industries
Supplier: SP Industries
LG-3861-104 LG-3861-102 LG-3861-110 LG-3861-108 LG-3861-100 LG-3861-106
76323-450EA 46.01 USD InStock
76323-450 76323-448 76323-446 76323-444 76323-442 76323-440
SP Wilmad-LabGlass Tall Form Weighing Bottles with Flat Top Cap, SP Industries
Bottles Weighing Bottles
Ideal for hygroscopic samples, these flat top, tall weighing bottles have an external ground joint that allows for sample transfer without interference to the seal.
  • Flat top permits stacking for increased desiccator capacity
  • High form shape decreases bottle footprint
  • Caps can be used as weighing dishes
  • Light weight allows simple tare settings
ASTM Type 1, Class A Borosilicate glass construction.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty