Would you like to log in again?
Stainless Steel Pipette Cans, Bellco Glass
Supplier: Bellco Glass
1217-11106
102681-542EA 73.64 USD InStock
102681-542
Stainless Steel Pipette Cans, Bellco Glass
Boxes Pipette Sterilization Containers
Heavy gauge rectangular construction for easy stacking and storage.
  • Silicone pads on inside of top and bottom protect pipette tips
  • Cans will accommodate up to 49 Pasteur pipettes (5.75" pipettes)
Can be sterilized by autoclave or dry heat (do not exceed 400 °F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty