Would you like to log in again?
Pipet-Aid™ Pipet-Aid XP, Bellco Glass
Supplier: Bellco Glass
1225-86000
100486-770CS 883.62 USD InStock
100486-770
Pipet-Aid™ Pipet-Aid XP, Bellco Glass
Pipettes Pipette Controllers
Rechargeable self-contained unit with removable stand.
  • Unit provides two hours of continuous pipetting without recharging
Three independent speed ranges can be set for both fill and dispense rates.
Delivery information: Includes battery charger and tissue culture nose piece.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty