Would you like to log in again?
Pyramid Brand Non Filtered Tips, Phenix Research Products
Supplier: Phenix
TS-200RF 1147C01 TS-200 TS-200BRL 1147B82 TS-200L TS-200BR 1147C03 TS-200BRLS TS-200RFL
101100-572PK 63.38 USD InStock
101100-572 101100-342 101100-760 101100-498 101100-478 101100-646 101100-668 101100-878 101100-416 101100-590
Pyramid Brand Non Filtered Tips, Phenix Research Products
Pipette Tips
These Pyramid brand pipette tips works with discovery pipettors, ovation pipettors, excel electronic pipettors and universal fit Gilson or Rainin style pipettors, except LTS style.
  • Universal fit
  • Certified RNase, DNase and pyrogen free
These are available in three different colors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty