Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crystal violet, HARLECO®, certified by the Biological Stain Commission for Gram staining, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Basic Violet 3GentiavermMethylrosanilide chlorideHexamethyl VioletVianinGentian violetMethyl violet 10BCI 42555Hexamethyl-p-rosaniline chlorideN-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chlorideDimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-yliden)ammoniumchloridViocid

192-12 192-16 192-12 192-29 192-16 192-29
EM-192-29EA 924.98 USD InStock
CA15204-287 EM-192-29 CA192-29 CA15204-120 15204-120 34172-132
Crystal violet, HARLECO®, certified by the Biological Stain Commission for Gram staining, Sigma-Aldrich®
Crystal Violet
Used for staining bacteria and spores by Gram’s method.


Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
Melting Pt: 189…194 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
IVDLogo
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty