Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogen carbonate 5% in aqueous solution for Gram staining

Supplier: Electron Microscopy Sciences
Synonyms: Sodium bicarbonate

26105-02
101411-030EA 39.71 USD InStock
101411-030
Sodium hydrogen carbonate 5% in aqueous solution for Gram staining
Sodium hydrogen carbonate
Reagent of Brown and Brenn Method for Gram Positive and Gram Negative Bacteria

Gram Negative Bacteria for bacteria stain: Gram + / Gram – bacteria and filaments of nocardia, etc

Stain results - Gram + Bacteria, Nocardia and Actinomyces Filaments: Blue, Gram – Bacteria, Nuclei: Red, Additional tissue elements: Yellow


Formula: NaHCO₃
MW: 84.01 g/mol
MDL Number: MFCD00003528
CAS Number: 144-55-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/81/16640581.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty