Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Dodecyl-β-D-maltoside ≥99.0%, high purity

Supplier: VWR
Synonyms: Dodecyl-β-D-maltopyranoside(2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triolN-Dodecyl-Beta-MaltosideLauryl-β-D-maltosideDDM

J424-1G
97063-172EA 195.32 USD InStock
97063-172
n-Dodecyl-β-D-maltoside ≥99.0%, high purity
n-Dodecyl-β-D-maltoside
Non-ionic - containing no charge (polyoxyethylenes or alkyl glucosides).
Non-ionic detergents are suited for solubilizing membrane proteins without altering biological activity.


Formula: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
MDL Number: MFCD00043012
CAS Number: 69227-93-6
Order Now
Specification Test Results
Abs.@225nm (1%, Water) ≤ 0.1
Abs.@260nm (1%, Water) ≤ 0.06
Abs.@280nm (1%, Water) ≤ 0.04
Abs.@340nm (1%, Water) ≤ 0.02
Alpha (HPLC) ≤ 15 %
pH (1%, Water) @25C 5--8
Purity ≥ 99.0 %
Solubility (1%, Water) PASS / Fail

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty