Would you like to log in again?
Laboratory Glassware Washer PG8536, Miele
Supplier: Miele
7831510
76311-456EA 40318.37 USD InStock
76311-456
Laboratory Glassware Washer PG8536, Miele
Glassware Washers
Laboratory glassware washer, fully programmable, HEPA forced-air drying, conductivity meter and two powerful pumps.
  • Highest level of flexibility – individually programmable controls
  • Security – spray arm monitoring and automatic mobile unit recognition
  • Perfect drying results – efficient hot air drying
Programmable controls – The programmable controls give the washer-disinfectors and thermal disinfectors a high level of flexibility. The machines are pre-equipped with numerous programs to cover the most diverse implementation areas. Authorized users can modify these programs or upload their own programs.
For reliable results – Outstanding reprocessing results are achieved through the spray arm monitoring, which immediately detects irregular spray pressure or any load blockages. In combination with the automatic mobile unit recognition, the highest level of safety is guaranteed. By using a magnetic strip, the washer-disinfector recognizes the different mobile unit types with their specific loads and automatically selects the appropriate reprocessing program.
Efficient drying – Active hot air drying is the optimum solution for hollow instruments or narrow-lumened laboratory glassware. The upstream H13 HEPA filter ensures optimum particle separation from the drying air. The filter is easily accessible via a side unit.
The PG 8536 Laboratory Glassware Washer is fully programmable, 158 gpm, with integrated liquid dispensing, and HEPA-filtered drying.
Certifications: EN 61010-1, EN 601010-1, CSA approved, CE-marked.
Ordering information: Accessories not included and must be ordered separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty