Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silica Magnetic Beads, G-Biosciences

Supplier: G-Biosciences
786-917 786-916 786-915
76285-568EA 811.49 USD InStock
76285-568 76285-566 76285-570
Silica Magnetic Beads, G-Biosciences
Beads Magnetic Beads
Silica magnetic beads are Fe3O₄ magnetic beads coated with a silicon dioxide (SiO₂) layer. Since silica is able to bind to the nucleic acids silica magnetic beads serve as a simple and efficient tool for plasmid DNA purification for transfection or sequencing applications, genomic DNA purification for research or clinical applications, RNA purification for qPCR analysis, or PCR product clean-up for downstream analysis.

  • Capacity: >4 mg DNA/ml beads
  • Particle size: 2.5 to 4.5 µm
  • Bead concentration: 50 mg/ml
  • Bead density: ~1.03 g/cm³
  • Simple, short procedure
  • No costly columns required
  • No time consuming centrifugations required

Applications include plasmid DNA isolation, genomic DNA isolation, RNA purification for qPCR and PCR product clean up.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty