Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safranine O, HARLECO®, certified by the Biological Stain Commission, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Safranin TSafranin Y3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chlorideSafranin OGossypimine3,7-Diamino-2,8-dimethyl-5-phenylphenazin-5-ium chlorideCI 50240Basic Red 2Cotton RedSafranin ASafranin

350-12 350-16 350-12 350-16
11214-844EA 410.23 USD InStock
CA11214-844 11214-844 34172-064 CA34172-064
Safranine O, HARLECO®, certified by the Biological Stain Commission, Sigma-Aldrich®
Safranine O
Used in various staining protocols such as Gram staining. Can be used as a cytological counterstain.


Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty