Would you like to log in again?
96-Well Multi-Tier™ Reaction Plates, Topas, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Multi-Tier™
CG-1915-21 CG-1915-20 CG-1915-32 CG-1915-03 CG-1915-24 CG-1915-31 CG-1915-02 CG-1915-22 CG-1915-01 CG-1915-30
77776-428EA 227.08 USD InStock
77776-428 77776-438 77776-426 77776-442 77776-430 77776-440 77776-424 77776-434 77776-444 77776-432
96-Well Multi-Tier™ Reaction Plates, Topas, Chemglass
Microplates Deep Well Microplates
The Multi-Tier™ 96 well reaction plates have a fixed height or can be multi-tiered to accept up to 1.5 ml borosilicate glass inserts.
  • Base plate and spacers are constructed of topas for greater chemical resistance and autoclaving
  • Borosilicate glass inserts provide excellent solvent resistance and thermal stability
  • Adjustable capacity glass wells: 0.5 to 1.5 ml
  • Standard 8×12 format alpha-numeric: A-H, 1-12
  • Base plate Is open permitting the glass to be in contact with heating or cooling media
  • Locking cover secures inserts in place
  • Conical design of glass inserts permits maximum sample recovery
  • Glass inserts are securely held in place permitting plates to be used with shakers/vortexers for agitation
  • Prescored PTFE/ silicone septa permits easier needle penetration
  • PTFE/Silicone stopper plugs prevent cross contamination and evaporation of individual reactions/samples
Plates have the same footprint as standard 96 well polystyrene plates and are compatible with 96 well automation workstations. Complete setups include: Base Plate, Glass Inserts, Prescored PTFE/Silicone Septa, and Snap On Cover.
Note: Plates are autoclavable.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty