Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Sulfosalicylic acid solution 3%, HARLECO® for microbiology, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 2-Hydroxy-5-sulphobenzoic acid2-Hydroxy-5-sulfobenzoic acid

1506A-75 1506A-75
34172-026EA 98.22 USD InStock
34172-026 CA34172-026
5-Sulfosalicylic acid solution 3%, HARLECO® for microbiology, Sigma-Aldrich®
5-Sulfosalicylic acid solutions
Used for semi-quantitative visual determination of albumin in urine by SSA precipitation method


Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00007508
CAS Number: 5965-83-3
Order Now
Specification Test Results
N/A N/A

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty