Would you like to log in again?
Metro Rack Back-Load Glove Dispensers, S-Curve
Supplier: S-CURVE TECHNOLOGIES
BLD-2400
82028-178CS 534.9 USD InStock
82028-178
Metro Rack Back-Load Glove Dispensers, S-Curve
Glove Dispensers and Glove Box Holders
Designed to fit a 24" deep Metro Rack. Dispenses product from the front while the rear opening allows restocking from behind the rack.
  • Designed to fit 3 dispensers on a 48" wide rack
  • Restock dispenser on the rack
  • Cannot be wall mounted
Picture is of the FLD-2400. The BLD-2400 is a similar design except it opens in the back.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty