Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® U Microbalances

Supplier: VWR
VWR®
VWR-5U VWR-11U VWR-2.1U VWR-31U VWR-2U VWR-21U VWR-6.1U
76321-874EA 32079.92 USD InStock
76321-874 76321-872 76321-866 76446-824 76446-826 76321-868 76321-870
VWR® U Microbalances
Balances and Scales
The VWR® U-Series Microbalances utilize advanced weighing technology allowing for the highest accuracy and repeatability in the smallest sample size.

  • Capacities from 2.1 to 31 g
  • Readabilites from 0.1 to 1 µg
  • Automatic level control and motorized glass draft shield
  • Hands-free operation via IR proximity sensors
  • Automatic, internal calibration mechanism
  • 5.7" color, resistive touch screen
  • 16 GB RAM to allow for expanded data management
  • VWR-2.1U and VWR-6.1U ultra microbalances include ADRFT-01 anti-draft chamber standard

Ideal for continuous monitoring of environmental conditions (temperature, humidity, pressure, and vibrations).

Three years warranty.

Certifications: Comply with USP conformatity regulations (Section 41 and 1251) to allow for unsurpassed quality of measurements. CE Marked.

Ordering information: Onsite installation and training of equipment included with purchase (continental United States and Canada). Please refer to related information tab for more information. To allow for the VWR® U-Series to perform to the highest standard, please refer to 'laboratory conditions' document with related information tab. Drop-shipment only.

Packaging: Packaged on small pallet.

Caution: Fragile. Handle with extreme care.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty